Skip to Content
Merck
All Photos(1)

Key Documents

W365203

Sigma-Aldrich

4-(p-Acetoxyphenyl)-2-butanone

≥96%, FG

Synonym(s):

4-(3-Oxobutyl)phenyl acetate, raspberry ketone acetate, 4-(4-Acetoxyphenyl)-2-butanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4CH2CH2COCH3
CAS Number:
Molecular Weight:
206.24
FEMA Number:
3652
Beilstein:
1961620
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.288
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥96%

refractive index

n20/D 1.509 (lit.)

bp

123-124 °C/0.2 mmHg (lit.)

density

1.099 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; blueberry; jam; raspberry

SMILES string

CC(=O)CCc1ccc(OC(C)=O)cc1

InChI

1S/C12H14O3/c1-9(13)3-4-11-5-7-12(8-6-11)15-10(2)14/h5-8H,3-4H2,1-2H3

InChI key

UMIKWXDGXDJQJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service