Skip to Content
Merck
All Photos(1)

Documents

W309508

Sigma-Aldrich

10-Undecenal

≥95%, FG, stabilized

Synonym(s):

Undecylenic aldehyde, 10-Undecenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CH(CH2)8CHO
CAS Number:
Molecular Weight:
168.28
FEMA Number:
3095
EC Number:
Council of Europe no.:
122
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.035

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

vapor density

>1 (vs air)

Assay

≥95%

contains

synthetic α-tocopherol as stabilizer

refractive index

n20/D 1.4427 (lit.)

bp

98-100 °C/3 mmHg (lit.)

density

0.84 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fatty; waxy; citrus; rose; sweet

storage temp.

2-8°C

SMILES string

[H]C(=O)CCCCCCCCC=C

InChI

1S/C11H20O/c1-2-3-4-5-6-7-8-9-10-11-12/h2,11H,1,3-10H2

InChI key

OFHHDSQXFXLTKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>140.0 °F - closed cup

Flash Point(C)

> 60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service