Key Documents
MT30027776
Mettler Toledo electrode cable
connector: K8SD, Mini-LTW, L 1.2 m
Synonym(s):
Mettler-Toledo electrode cable
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
Pricing and availability is not currently available.
manufacturer/tradename
Mettler-Toledo 30027776
L
1.2 m
General description
Dedicated cable to connect InLab® Smart Pro-ISM to SevenCompact S220, K8SD to Mini-LTW, 1.2 m
Mettler Toledo electrodes can easily be connected to various third-party instruments. All you have to do is select the appropriate cable. We offer a wide choice of cable lengths and plug combinations. For Mettler Toledo reference electrodes and temperature probes.
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)