Documents
BAH367150000
SP Bel-Art® Quaver Vibrating Spatula
L 7 in. (17.78 cm), brown FEP blade
Synonym(s):
spoon
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
brown FEP blade
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H36715-0000
L
7 in. (17.78 cm)
General description
Make accurate additions of precious materials as needed. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving you control over transfer from spatula to weighing dish.
Features and Benefits
  • Available with a plain stainless steel blade or with a brown FEP coating
  • Coating complies with FDA regulations governing direct food contact applications; reduces transfer losses
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP