Skip to Content
Merck
All Photos(2)

Key Documents

912018

Sigma-Aldrich

7-Methyl-2-phenylimidazo[1,2-a]pyrazin-3(7H)-one

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11N3O
CAS Number:
Molecular Weight:
225.25
UNSPSC Code:
12352200

Assay

≥95%

form

powder

storage temp.

2-8°C

InChI

1S/C13H11N3O/c1-15-7-8-16-11(9-15)14-12(13(16)17)10-5-3-2-4-6-10/h2-9H,1H3

InChI key

DGQHNBJMAAENRX-UHFFFAOYSA-N

Application

7-Methyl-2-phenylimidazo[1,2-a]pyrazin-3(7H)-one is a hydrogen bonding sensor developed by the Kozlowski group. This colormetric probe has been used to assess the relative reactivity of hydrogen bonding organocatalysts as well as to measure hydrogen bond strength in pharamceutically relevant molecules.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Fan et al.
The Journal of organic chemistry, 83(18), 10855-10863 (2018-07-20)
Charged thiourea derivatives containing one and two methylated or octylated pyridinium ion centers and a tetraarylborate or triflate counteranion are reported. These novel catalysts are much more active in the Friedel-Crafts reactions of trans-β-nitroalkenes with N-methylindoles than the privileged N
Phuong N H Huynh et al.
Journal of the American Chemical Society, 134(38), 15621-15623 (2012-09-15)
A sensor has been developed to quickly and simply assess the relative reactivity of different hydrogen-bonding catalysts. Specifically, blue-shifts seen upon treatment of H-bonding catalysts with the colorimetric compound 7-methyl-2-phenylimidazo[1,2-a]pyrazin-3(7H)-one correlate well to the K(eq) of binding to the sensor.
Ryan R Walvoord et al.
Journal of the American Chemical Society, 136(45), 16055-16065 (2014-10-18)
A spectrophotometric sensor is described that provides a useful assessment of the LUMO-lowering provided by catalysts in Diels-Alder and Friedel-Crafts reactions. A broad range of 33 hydrogen-bonding catalysts was assessed with the sensor, and the relative rates in the above

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service