Skip to Content
Merck
All Photos(2)

Key Documents

900998

Sigma-Aldrich

Dipyridin-4-ylmethanone

≥95%

Synonym(s):

Bis(4-pyridyl) ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8N2O
CAS Number:
Molecular Weight:
184.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

mp

142 °C

functional group

ketone

SMILES string

O=C(C1=CC=NC=C1)C2=CC=NC=C2

InChI

1S/C11H8N2O/c14-11(9-1-5-12-6-2-9)10-3-7-13-8-4-10/h1-8H

InChI key

CLAOXGJAXHTMPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dipyridin-4-ylmethanone may be used as a precursor to synthesize photoresponsive ligands such as diazodi(4-pyridyl)methane, which can be used in metal-carbene hetero-spin systems. This same product was previously listed as CBR00084.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diazodi(4-pyridyl)methane and Diazophenyl-(4-pyridyl)methane as Photoresponsive Ligands for Metal?Carbene Hetero-Spin Systems.
K Noboru, et al.
Angewandte Chemie (International Edition in English), 35, 755-757 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service