Skip to Content
Merck
All Photos(1)

Key Documents

792713

Sigma-Aldrich

2-Bromo-4-(trifluoromethyl)benzenesulfonyl fluoride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3BrF4O2S
CAS Number:
Molecular Weight:
307.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

65-70 °C

functional group

bromo
fluoro

SMILES string

O=S(C1=C(Br)C=C(C(F)(F)F)C=C1)(F)=O

InChI

1S/C7H3BrF4O2S/c8-5-3-4(7(9,10)11)1-2-6(5)15(12,13)14/h1-3H

InChI key

DVDPGVYCZZAPAK-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service