Skip to Content
Merck
All Photos(1)

Key Documents

779733

Sigma-Aldrich

1-Bromo-2-(bromomethyl)naphthalene

≥96.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8Br2
CAS Number:
Molecular Weight:
299.99
Beilstein:
1943405
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (GC)
96.0-104.0% (with AgNO3 after O2 combustion)

form

powder

SMILES string

BrCc1ccc2ccccc2c1Br

InChI

1S/C11H8Br2/c12-7-9-6-5-8-3-1-2-4-10(8)11(9)13/h1-6H,7H2

InChI key

DQTOCXIHYIQHCK-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service