Skip to Content
Merck
All Photos(1)

Key Documents

778621

Sigma-Aldrich

2,3,4,6-Tetra-O-pivaloyl-α-D-glucopyranosyl bromide

≥90% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H43BrO9
CAS Number:
Molecular Weight:
579.52
Beilstein:
3576922
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥90% (TLC)

form

powder

optical activity

[α]/D 152.0±3.0°, c = 3 in chloroform

storage temp.

2-8°C

SMILES string

CC(C)(C)C(=O)OC[C@H]1O[C@H](Br)[C@H](OC(=O)C(C)(C)C)[C@@H](OC(=O)C(C)(C)C)[C@@H]1OC(=O)C(C)(C)C

InChI

1S/C26H43BrO9/c1-23(2,3)19(28)32-13-14-15(34-20(29)24(4,5)6)16(35-21(30)25(7,8)9)17(18(27)33-14)36-22(31)26(10,11)12/h14-18H,13H2,1-12H3/t14-,15-,16+,17-,18+/m1/s1

InChI key

BSDBCYHGMPHOAL-SFFUCWETSA-N

Application

Königs-Knorr glycosyl donor with considerably lower tendency to form orthoester side products during glycosylation than the traditional acetobromoglucose

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Presser et al.
Monatshefte fur Chemie / Chemical Monthly, 137, 365-365 (2006)
H. Kunz and A. Harreus
Liebigs Ann. Chem., 41-48 (1982)
H. Kunz and A. Harreus
Liebigs Ann. Chem., 717-730 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service