Skip to Content
Merck
All Photos(2)

Key Documents

775673

Sigma-Aldrich

Methyl 4,7,8,9-tetra-O-acetyl-N-acetylneuraminate

95%

Synonym(s):

4,7,8,9-Tetra-O-acetyl-N-acetylneuraminic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29NO13
CAS Number:
Molecular Weight:
491.44
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

185-189 °C

storage temp.

2-8°C

SMILES string

COC(=O)[C@]1(O)C[C@H](OC(C)=O)[C@@H](NC(C)=O)[C@@H](O1)[C@H](OC(C)=O)[C@@H](COC(C)=O)OC(C)=O

InChI

1S/C20H29NO13/c1-9(22)21-16-14(31-11(3)24)7-20(28,19(27)29-6)34-18(16)17(33-13(5)26)15(32-12(4)25)8-30-10(2)23/h14-18,28H,7-8H2,1-6H3,(H,21,22)/t14-,15+,16+,17+,18+,20-/m0/s1

InChI key

CAKDMYCBPKFKCV-DCDDNHAVSA-N

Application

Versatile acetylated neuraminic acid or sialic acid derivative. N-acetylneuraminic acid is often present as the terminal sugar of glycoproteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kelm et al.
International review of cytology, 175, 137-240 (1997-01-01)
Sialic acids (Sias) are terminal components of many glycoproteins and glycolipids especially of higher animals. In this exposed position they contribute significantly to the structural properties of these molecules, both in solution and on cell surfaces. Therefore, it is not
S Narayanan
Annals of clinical and laboratory science, 24(4), 376-384 (1994-07-01)
The term sialic acid is used to describe derivatives of neuraminic acid, where the amino group of neuraminic acid is substituted by either an acetyl or glycolyl group. The unique structural features of the molecule, which includes a negative charge

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service