768464
2,7-Dihexylbenzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetrone
Synonym(s):
N,N′-(1-Hexyl)-1,4,5,8-naphthalenetetracarboxydiamide, NDI-C6
About This Item
Recommended Products
Assay
≥97.6%
form
powder
mp
205-210 °C
λmax
380, 360, 342 nm in dichloromethane (lit.)
semiconductor properties
N-type (mobility=0.7 cm2/V·s)
SMILES string
CCCCCCN1C(=O)c2ccc3C(=O)N(CCCCCC)C(=O)c4ccc(C1=O)c2c34
InChI
1S/C26H30N2O4/c1-3-5-7-9-15-27-23(29)17-11-13-19-22-20(14-12-18(21(17)22)24(27)30)26(32)28(25(19)31)16-10-8-6-4-2/h11-14H,3-10,15-16H2,1-2H3
InChI key
QGDZMDFMUJURJD-UHFFFAOYSA-N
General description
Application
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.
Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.
Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service