Skip to Content
Merck
All Photos(1)

Documents

752827

Sigma-Aldrich

Tin(IV) bromide

99.999% trace metals basis

Synonym(s):

Stannic bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnBr4
CAS Number:
Molecular Weight:
438.33
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.999% trace metals basis

bp

202 °C (lit.)

mp

31 °C (lit.)

density

3.34 g/mL at 25 °C (lit.)

SMILES string

Br[Sn](Br)(Br)Br

InChI

1S/4BrH.Sn/h4*1H;/q;;;;+4/p-4

InChI key

LTSUHJWLSNQKIP-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Used as a precursor for SnO2 through laser-assisted CVD with higher efficiency than other precursors such as dibutyl tin diacetate.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pettinari, C.; et al.
Inorgorganica Chimica Acta, 325, 103-103 (2001)
Hossenlopp; J.M.; et al.
Applied Organometallic Chemistry, 12, 147-147 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service