Skip to Content
Merck
All Photos(1)

Documents

747882

Sigma-Aldrich

4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid

95%

Synonym(s):

4-Bromo-1,3-dimethylpyrazole-5-carboxylic acid, 4-Bromo-2,5-dimethyl-2H-pyrazole-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7BrN2O2
CAS Number:
Molecular Weight:
219.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

powder

mp

237-242 °C

SMILES string

OC(C1=C(Br)C(C)=NN1C)=O

InChI

1S/C6H7BrN2O2/c1-3-4(7)5(6(10)11)9(2)8-3/h1-2H3,(H,10,11)

InChI key

ZUEAEVZRYGUFPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid (IUPAC name: methyl 4-bromo-2,5-dimethylpyrazole-3-carboxylate) is a pyrazole derivative. .

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service