Skip to Content
Merck
All Photos(1)

Key Documents

742236

Sigma-Aldrich

[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium triflate

≥98% (HPLC)

Synonym(s):

[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15F6IO3S
CAS Number:
Molecular Weight:
540.26
Beilstein:
20424731
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (HPLC)

form

solid

composition

carbon, 36.8-38.7% CH

SMILES string

CC1=CC(C)=C([I+]C2=CC=C(C=C2)C(F)(F)F)C(C)=C1.O=S([O-])(C(F)(F)F)=O

InChI

1S/C16H15F3I.CHF3O3S/c1-10-8-11(2)15(12(3)9-10)20-14-6-4-13(5-7-14)16(17,18)19;2-1(3,4)8(5,6)7/h4-9H,1-3H3;(H,5,6,7)/q+1;/p-1

InChI key

XTTMIKRDUJQRGJ-UHFFFAOYSA-M

Application

[4-(Trifluoromethyl)phenyl](2,4,6-trimethylphenyl)iodonium triflate can be used as:
  • A Lewis acid catalyst in the metal-free cationic polymerization of styrene and alkyl vinyl ether derivatives.
  • An arylating agent in the arylalkynylation of alkenes.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-Free Living Cationic Polymerization Using Diaryliodonium Salts as Organic Lewis Acid Catalysts
Haraguchi R, et al.
Macromolecules, 53, 4185?4192-4185?4192 (2020)
Copper-catalyzed enantioselective arylalkynylation of alkenes
Lei G, et al.
Chemical Science, 11(6), 1623-1628 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service