Skip to Content
Merck
All Photos(1)

Key Documents

736910

Sigma-Aldrich

3-Fluoro-2-iodoanisole

97%

Synonym(s):

1-Fluoro-2-iodo-3-methoxybenzene, 6-Fluoro-2-methoxyiodobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6FIO
CAS Number:
Molecular Weight:
252.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Assay

97%

form

liquid

refractive index

n20/D 1.597

density

1.859 g/mL at 25 °C

functional group

fluoro
iodo

SMILES string

COc1cccc(F)c1I

InChI

1S/C7H6FIO/c1-10-6-4-2-3-5(8)7(6)9/h2-4H,1H3

InChI key

CPTMVKLYWHEOMG-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service