Skip to Content
Merck
All Photos(1)

Documents

708712

Sigma-Aldrich

2,6-Dichloropyridine-4-carbonyl chloride

97%

Synonym(s):

2,6-Dichloroisonicotinoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl3NO
CAS Number:
Molecular Weight:
210.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.579

bp

243 °C

mp

25 °C

density

1.537 g/mL at 25 °C

InChI

1S/C6H2Cl3NO/c7-4-1-3(6(9)11)2-5(8)10-4/h1-2H

InChI key

QLRUROKTVFUQIV-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service