Skip to Content
Merck
All Photos(1)

Key Documents

70869

Sigma-Aldrich

1-Butyl-2,3-dimethylimidazolium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17F6N2P
CAS Number:
Molecular Weight:
298.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (HPLC)
≥97.0%

form

liquid

density

1.345 g/mL at 20 °C (lit.)

SMILES string

F[P-](F)(F)(F)(F)F.CCCCn1cc[n+](C)c1C

InChI

1S/C9H17N2.F6P/c1-4-5-6-11-8-7-10(3)9(11)2;1-7(2,3,4,5)6/h7-8H,4-6H2,1-3H3;/q+1;-1

InChI key

JWFPQAXAGSAKRF-UHFFFAOYSA-N

Application

1-Butyl-2,3-dimethylimidazolium hexafluorophosphate is an ionic liquid that can be used:
  • As a green solvent in the separation of endosulfan from water.[1]
  • As a binder in the preparation of carbon paste electrode, applicable in the detection of adenosine-5′-triphosphate in injection samples.[2]
  • As a component of the biphasic medium along with toluene, used in norbornene ring-opening metathesis polymerization.[3]

Physical form

ionic liquid

Other Notes

Ionic liquid suitable for use in strongly basic conditions, e.g. aldol condensation and metathesis reactions[4][5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitive Electrochemical Determination of Adenosine-5′-triphosphate with 1-Butyl-2, 3-dimethylimidazolium Hexafluorophosphate Based Carbon Paste Electrode
Wei L, et al.
International Journal of Electrochemical Science, 14(11), 10070-10078 (2019)
1-Butyl-2, 3-dimethylimidazolium hexafluorophosphate as a green solvent for the extraction of endosulfan from aqueous solution using supported liquid membrane
Pilli SR, et al.
Chemical Engineering Journal, 257, 56-65 (2014)
First ring-opening metathesis polymerization in an ionic liquid. Efficient recycling of a catalyst generated from a cationic ruthenium allenylidene complex
Csihony S, et al.
New. J. Chem., 26(11), 1667-1670 (2002)
Priya A Thomas et al.
International journal of molecular sciences, 12(6), 3989-3997 (2011-07-13)
The self-metathesis of methyl oleate and methyl ricinoleate was carried out in the presence of ruthenium alkylidene catalysts 1-4 in [bmim] and [bdmim][X] type ionic liquids (RTILs) (X = PF(6) (-), BF(4) (-) and NTf(2) (-)) using the gas chromatographic
S. Csihony et al.
New. J. Chem., 26, 1667-1667 (2002)

Articles

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service