Skip to Content
Merck
All Photos(3)

Key Documents

683825

Sigma-Aldrich

4-Fluoro-3-formylphenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6BFO3
CAS Number:
Molecular Weight:
167.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

215-220 °C

functional group

aldehyde
fluoro

storage temp.

2-8°C

SMILES string

OB(O)c1ccc(F)c(C=O)c1

InChI

1S/C7H6BFO3/c9-7-2-1-6(8(11)12)3-5(7)4-10/h1-4,11-12H

InChI key

YABSTJQEBSKPCG-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of arylmethylpyrrolidinylmethanols and amine derivatives via reaction with MIDA, followed by Suzuki reaction with halides or amination with amines
  • Preparation of N-(biarylsulfonyl) a-amino acids as MMP-12 inhibitors using a genetic algorithm
  • Preparation of para-substituted formylarylboronic esters for regioselective Suzuki-Miyaura cross-coupling with bromobenzene

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service