Skip to Content
Merck
All Photos(2)

Documents

669415

Sigma-Aldrich

2-Bromo-2′,6′-dichloro-3′-fluoroacetophenone

95%

Synonym(s):

2′,6′-Dichloro-3′-fluorophenacyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4BrCl2FO
Molecular Weight:
285.93
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

38-44 °C

storage temp.

2-8°C

SMILES string

Fc1ccc(Cl)c(c1Cl)C(=O)CBr

InChI

1S/C8H4BrCl2FO/c9-3-6(13)7-4(10)1-2-5(12)8(7)11/h1-2H,3H2

InChI key

QFVUFUXFUVITSO-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service