Skip to Content
Merck
All Photos(2)

Documents

597406

Sigma-Aldrich

2,4-Difluoro-3-formylphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H2(CHO)B(OH)2
CAS Number:
Molecular Weight:
185.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

mp

206-210 °C (lit.)

SMILES string

[H]C(=O)c1c(F)ccc(B(O)O)c1F

InChI

1S/C7H5BF2O3/c9-6-2-1-5(8(12)13)7(10)4(6)3-11/h1-3,12-13H

InChI key

PVAWONNALJEQJH-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daojin Li et al.
The Analyst, 142(23), 4494-4502 (2017-11-03)
Boronate affinity materials, as efficient sorbents for extraction, separation and enrichment of glycoproteins, have attracted more and more attention in recent years. However, conventional boronate affinity materials suffer from low binding affinity due to the use of single boronic acids
Daojin Li et al.
Journal of chromatography. A, 1384, 88-96 (2015-02-02)
Boronate affinity materials, as effective sample enrichment sorbents for glycoproteomic analysis, have attracted increasing attention in recent years. However, most of boronate affinity materials suffer from an apparent limitation, limited binding strength. As a result, extraction of glycoproteins of trace

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service