Recommended Products
Assay
97%
form
solid
mp
175-179 °C (lit.)
SMILES string
Oc1ncc(cc1[N+]([O-])=O)[N+]([O-])=O
InChI
1S/C5H3N3O5/c9-5-4(8(12)13)1-3(2-6-5)7(10)11/h1-2H,(H,6,9)
InChI key
KSZZJOXNRQKULB-UHFFFAOYSA-N
General description
2-hydroxy-3,5-dinitropyridine is an industrially important nitropyridine derivative. Its lead salt finds application in solid propellants as catalysts.
Application
2-hydroxy-3,5-dinitropyridine may be used in the preparation of bis([μ]-3,5-dinitropyridin-2-olato)bis[tetraaqua(3,5-dinitropyridin-2-olato)barium(II)].
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Constant-volume combustion energy of the lead salts of 2HDNPPb and 4HDNPPb and their effect on combustion characteristics of RDX-CMDB propellant.
Journal of Thermal Analysis and Calorimetry, 85(3), 791-794 (2006)
Bis (?-3, 5-dinitropyridin-2-olato) bis [tetraaqua (3, 5-dinitropyridin-2-olato) barium (II)].
Acta Crystallographica Section E, Structure Reports Online, 63(5), m1302-m1304 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service