549673
4′-Acetamido-2′-methylacetophenone
97%
Synonym(s):
N-(4-Acetyl-3-methylphenyl)acetamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CONHC6H3(CH3)COCH3
CAS Number:
Molecular Weight:
191.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
mp
137-140 °C (lit.)
SMILES string
CC(=O)Nc1ccc(C(C)=O)c(C)c1
InChI
1S/C11H13NO2/c1-7-6-10(12-9(3)14)4-5-11(7)8(2)13/h4-6H,1-3H3,(H,12,14)
InChI key
PTARWPNYVATTDE-UHFFFAOYSA-N
General description
4′-Acetamido-2′-methylacetophenone (4-acetamido-2-methylacetophenone) can be prepared by treating m-methylacetanilide with acetyl chloride in the presence of aluminum chloride in carbon disulfide.
Application
4′-Acetamido-2′-methylacetophenone may be used in the preparation of 4-amino-2-methylacetophenone by acid hydrolysis.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Estrogenic Biphenyls. IV. 3'-Alkyl-4-methoxybiphenyl-4'-carboxylic Acids.
Sato T and Oki M.
Bulletin of the Chemical Society of Japan, 30(9), 958-961 (1957)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service