Skip to Content
Merck
All Photos(1)

Key Documents

513644

Sigma-Aldrich

tert-Butylperoxy isopropyl carbonate solution

75 wt. % in odorless mineral spirits

Synonym(s):

Luperox® TBIC-M75

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCO3C(CH3)3
CAS Number:
Molecular Weight:
176.21
UNSPSC Code:
12162002

concentration

75 wt. % in odorless mineral spirits

refractive index

n20/D 1.409

density

0.866 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Polymerization initiator.

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service