Skip to Content
Merck
All Photos(1)

Key Documents

497819

Sigma-Aldrich

5-Chloropentylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)5ZnBr
CAS Number:
Molecular Weight:
250.88
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

concentration

0.5 M in THF

density

0.956 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

ClCCCCC[Zn]Br

InChI

1S/C5H10Cl.BrH.Zn/c1-2-3-4-5-6;;/h1-5H2;1H;/q;;+1/p-1

InChI key

SOVQGFDZTBAGEL-UHFFFAOYSA-M

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-6.2 °F - (THF)

Flash Point(C)

-21.2 °C - (THF)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service