Skip to Content
Merck
All Photos(1)

Key Documents

495298

Sigma-Aldrich

1,8-Naphthalenedimethanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CH2OH)2
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

153-156 °C (lit.)

functional group

hydroxyl

SMILES string

OCc1cccc2cccc(CO)c12

InChI

1S/C12H12O2/c13-7-10-5-1-3-9-4-2-6-11(8-14)12(9)10/h1-6,13-14H,7-8H2

InChI key

DINZUYYYXDLSJE-UHFFFAOYSA-N

General description

1,8-Naphthalenedimethanol, also known as 1,8-bis(hydroxymethyl)naphthalene, is an aromatic diol. It can be synthesized by the reduction of 1,8-naphthalic anhydride using lithium aluminum hydride. It crystallizes in the monoclinic space group P21/n.

Application

1,8-Naphthalenedimethanol may be used in the synthesis of 1,8-bis-(chloromethyl)-naphthalene and 1,8-dimethylnaphthalene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mild and Efficient Synthesis of 1,8-Naphthalide and 1,8-Naphthalenedimethanol.
Luo H, et al.
Synthetic Communications, 34(12), 2269-2275 (2004)
The Alkaloids of Delphinium Barbeyi H.
Cook WB and Beath OA.
Journal of the American Chemical Society, 74(6), 1411-1415 (1952)
G D Bennett et al.
Acta crystallographica. Section C, Crystal structure communications, 49 ( Pt 1), 64-68 (1993-01-15)
1,8-Naphthalenedimethanol, C12H12O2, M(r) = 188.24, monoclinic, P2(1)/n, a = 8.549(4), b = 4.856(3), c = 22.604(4) A, beta = 94.24(3) degrees, V = 935.8(7) A 3, Z = 4, Dx = 1.34 g cm-3, lambda(Mo K alpha) = 0.71073 A
A Synthesis of Pleiadiene and Acepleiadylene1.
Boekelheide V and Vick GK.
Journal of the American Chemical Society, 78(3), 653-658 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service