Skip to Content
Merck
All Photos(1)

Key Documents

466956

Sigma-Aldrich

4-Fluorophenylacetyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2COCl
CAS Number:
Molecular Weight:
172.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.512 (lit.)

bp

60 °C/8 mmHg (lit.)

density

1.259 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1ccc(CC(Cl)=O)cc1

InChI

1S/C8H6ClFO/c9-8(11)5-6-1-3-7(10)4-2-6/h1-4H,5H2

InChI key

SIOJFYRPBYGHOO-UHFFFAOYSA-N

Application

4-Fluorophenylacetyl chloride may be used in the preparation of bicyclic pyrazolones[1][2] and dihydropyrrol-2-ones.[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of orally bioavailable bicyclic pyrazolones as inhibitors of tumor necrosis factor-a production.
Clark MP, et al.
Journal of Medicinal Chemistry, 47(11), 2724-2727 (2004)
Drug Des. Discovery, 9, 155-155 (1992)
Convergent synthesis of 2, 3-bisarylpyrazolones through cyclization of bisacylated pyrazolidines and hydrazines.
Brugel TA, et al.
Tetrahedron Letters, 47(19), 3195-3198 (2006)
Drug Des. Discovery, 11, 115-115 (1994)
Synthesis of phenylquinoxaline oligomers containing pendant electron donating and withdrawing groups.
Baek J-B, et al.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 47(1), 341-341 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service