Skip to Content
Merck
All Photos(1)

Key Documents

405833

Sigma-Aldrich

(3,5-Dimethoxyphenyl)acetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2CO2H
CAS Number:
Molecular Weight:
196.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

102-103 °C (lit.)

SMILES string

COc1cc(CC(O)=O)cc(OC)c1

InChI

1S/C10H12O4/c1-13-8-3-7(5-10(11)12)4-9(6-8)14-2/h3-4,6H,5H2,1-2H3,(H,11,12)

InChI key

FFPAFDDLAGTGPQ-UHFFFAOYSA-N

General description

(3,5-Dimethoxyphenyl)acetic acid is a carboxylic acid. Its physical properties like density, freezing point, boiling point and refractive index have been determined.[1]

Application

(3,5-Dimethoxyphenyl)acetic acid may be employed in a study to investigate the isolation of phananthrene derivatives from the heartwood of Combretum hereroense.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chemical constituents of the combretaceae. Part IV. Phenanthrene derivatives from the heartwood of Combretum hereroense.
Llewellyn R M.
Journal of the Chemical Society. Perkin Transactions 1, 1179-1181 (1973)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 286-286 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service