Skip to Content
Merck
All Photos(1)

Key Documents

371440

Sigma-Aldrich

(1-tert-Butylvinyloxy)trimethylsilane

98%

Synonym(s):

(2,2-Dimethyl-1-methylenepropoxy)trimethylsilane, 3,3-Dimethyl-2-trimethylsiloxy-1-butene, Pinacolone enol trimethylsilyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC(=CH2)OSi(CH3)3
CAS Number:
Molecular Weight:
172.34
Beilstein:
2204704
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.409 (lit.)

bp

140-142 °C (lit.)

density

0.798 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C(=C)O[Si](C)(C)C

InChI

1S/C9H20OSi/c1-8(9(2,3)4)10-11(5,6)7/h1H2,2-7H3

InChI key

PEHJULPJEVIIFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1-tert-Butylvinyloxy)trimethylsilane is a silyl enol. It belongs to organosilicone class of compounds. Enthalpy of vaporization of (1-tert-Butylvinyloxy)trimethylsilane at boiling point has been reported.[1]

Application

(1-tert-Butylvinyloxy)trimethylsilane ((2,2-Dimethyl-1-methylenepropoxy)trimethylsilane) may be used in the synthesis of cis-3,3-dimethyl-1-(6-phenethyl-3,6-dihydro-2H-pyran-2-yl)butan-2-one.[2]
(1-tert-Butylvinyloxy)trimethylsilane may be used in the preparation of 1-(4-bromophenyl)-4,4-dimethylpentan-3-one.[3]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 574-574 (2014)
Salvatore Ferla et al.
Journal of medicinal chemistry, 57(18), 7702-7715 (2014-08-26)
The synthesis of imidazole styrylbenzamide, tert-butyl styrylimidazole, and tert-butyl styrylsulfonate derivatives is described. Evaluation of binding affinity and inhibitory activity against CYP24A1 identified the imidazole styrylbenzamides as potent inhibitors of CYP24A1, having selectivity with respect to CYP27B1 comparable with or
Robert J Hinkle et al.
Tetrahedron, 65(34), 6834-6839 (2010-02-18)
The rapid synthesis of cis-2,6-disubstituted dihydropyrans is achieved in a three-component, one-pot cascade reaction. BiBr(3)-ediated addition of ketene silyl acetals or silyl enol ethers to beta,gamma-unsaturated cis-4-trimethylsilyl-3-butenal provides a Mukaiyama aldol adduct containing a vinylsilane moiety tethered to a silyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service