Skip to Content
Merck
All Photos(1)

Key Documents

310786

Sigma-Aldrich

Benzo[f]ninhydrin monohydrate

97%

Synonym(s):

2,2-Dihydroxy-(1H)-benz[f]indene-1,3(2H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H6O3 · H2O
CAS Number:
Molecular Weight:
228.20
Beilstein:
2459087
MDL number:
UNSPSC Code:
12352200

Assay

97%

mp

150 °C (dec.) (lit.)

SMILES string

OC1(O)C(=O)c2cc3ccccc3cc2C1=O

InChI

1S/C13H8O4/c14-11-9-5-7-3-1-2-4-8(7)6-10(9)12(15)13(11,16)17/h1-6,16-17H

InChI key

YEKNOJJZNVJMMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service