Skip to Content
Merck
All Photos(1)

Key Documents

260207

Sigma-Aldrich

1,2-Epoxydodecane

90%

Synonym(s):

1-Dodecene oxide, Decenoxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O
CAS Number:
Molecular Weight:
184.32
Beilstein:
105569
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

2 mmHg ( 100 °C)

Assay

90%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

124-125 °C/15 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCC1CO1

InChI

1S/C12H24O/c1-2-3-4-5-6-7-8-9-10-12-11-13-12/h12H,2-11H2,1H3

InChI key

MPGABYXKKCLIRW-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meiling Zhou et al.
Journal of controlled release : official journal of the Controlled Release Society, 310, 24-35 (2019-08-14)
The lack of effective glioma therapeutics mandates the development of novel treatment strategies. Hepatoma-derived growth factor (HDGF) has been considered as a potential glioma therapeutic target, and its expression level in gliomas is positively related to the malignant grade. Although
Christopher J Backlund et al.
Acta biomaterialia, 29, 198-205 (2015-10-20)
The effect of nitric oxide (NO)-releasing dendrimer hydrophobicity on Streptococcus mutans killing and biofilm disruption was examined at pH 7.4 and 6.4, the latter relevant to dental caries. Generation 1 (G1) poly(amidoamine) (PAMAM) dendrimers were modified with alkyl epoxides to
Anna K Blakney et al.
Gene therapy, 26(9), 363-372 (2019-07-14)
Self-amplifying RNA (saRNA) is a promising biotherapeutic tool that has been used as a vaccine against both infectious diseases and cancer. saRNA has been shown to induce protein expression for up to 60 days and elicit immune responses with lower

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service