Skip to Content
Merck
All Photos(2)

Key Documents

246697

Sigma-Aldrich

1-Amino-4-bromo-2-methylanthraquinone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10BrNO2
CAS Number:
Molecular Weight:
316.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

245 °C (dec.) (lit.)

SMILES string

Cc1cc(Br)c2C(=O)c3ccccc3C(=O)c2c1N

InChI

1S/C15H10BrNO2/c1-7-6-10(16)11-12(13(7)17)15(19)9-5-3-2-4-8(9)14(11)18/h2-6H,17H2,1H3

InChI key

VIQMJMDPUIBXQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Amino-4-bromo-2-methylanthraquinone has been used in the synthesis of 4-substituted 1-amino-2-methylanthraquinone derivatives:
  • 1-amino-2-methyl-4-phenylaminoanthraquinone[1]
  • 1-amino-2-methyl-4-(3-methylphenylamino)anthraquinone[1]
  • 1-amino-2-methyl-4-(4-methylphenylamino)anthraquinone[1]
  • 1-amino-2-methyl-4-(1-naphthylamino)anthraquinone[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Younis Baqi et al.
Purinergic signalling, 5(1), 91-106 (2008-06-06)
Reactive blue 2 (RB-2) had been characterized as a relatively potent ectonucleoside triphosphate diphosphohydrolase (E-NTPDase) inhibitor with some selectivity for NTPDase3. In search for the pharmacophore and to analyze structure-activity relationships we synthesized a series of truncated derivatives and analogs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service