Skip to Content
Merck
All Photos(1)

Key Documents

223018

Sigma-Aldrich

Retinoic acid

98%

Synonym(s):

ATRA, Tretinoin, Vitamin A acid, all-trans-Retinoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
Beilstein:
2057223
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

Assay

98%

mp

180-181 °C (lit.)

SMILES string

CC1=C(\C=C\C(C)=C\C=C\C(C)=C\C(O)=O)C(C)(C)CCC1

InChI

1S/C20H28O2/c1-15(8-6-9-16(2)14-19(21)22)11-12-18-17(3)10-7-13-20(18,4)5/h6,8-9,11-12,14H,7,10,13H2,1-5H3,(H,21,22)/b9-6+,12-11+,15-8+,16-14+

InChI key

SHGAZHPCJJPHSC-YCNIQYBTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Antitumor agent[1] and inducing factor in leukemia differentiation.[2]

Biochem/physiol Actions

all−trans−Retinoic acid (ATRA) is a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR). The bound RAR and RXR act as transcription factors that regulate the growth and differentiation of both normal and malignant cells. Cytochromes P450 (CYPs) catalyze the 4-hydroxylation of ATRA. Retinoic acid primes embryonic stem cells to become neurons.
Ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR); primes embryonic stem cells to become neurons.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H M Beere et al.
Anti-cancer drug design, 8(4), 299-322 (1993-08-01)
Differentiation therapy focuses on the development and use of specific agents designed to selectively engage the process of terminal differentiation, leading to the eventual elimination of tumorigenic cells and rebalance of normal cellular homeostasis. Extensive in vitro study of the
Cellular and Molecular Biology, 40, 297-297 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service