Skip to Content
Merck
All Photos(1)

Key Documents

130796

Sigma-Aldrich

3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)-2-naphthalenecarboxylic acid

indicator grade, Dye content 60 %

Synonym(s):

2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthoic acid, Cal-Red, Calconcarboxylic acid, HHSNN, Patton and Reeder’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H5[N=NC10H5(OH)SO3H]CO2H
CAS Number:
Molecular Weight:
438.41
Beilstein:
727492
EC Number:
MDL number:
UNSPSC Code:
12171500
Pricing and availability is not currently available.

grade

indicator grade

composition

Dye content, 60%

mp

300 °C (lit.)

λmax

366 nm
560 nm (2nd)

SMILES string

OC(=O)c1cc2ccccc2c(N=Nc3c(O)cc(c4ccccc34)S(O)(=O)=O)c1O

Legal Information

Cal-Red is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service