Merck
  • Neonatal exposure to low doses of endosulfan induces implantation failure and disrupts uterine functional differentiation at the pre-implantation period in rats.
Neonatal exposure to low doses of endosulfan induces implantation failure and disrupts uterine functional differentiation at the pre-implantation period in rats.
Molecular and cellular endocrinology (2014-12-09)
María M Milesi, Ramiro Alarcón, Jorge G Ramos, Mónica Muñoz-de-Toro, Enrique H Luque, Jorgelina Varayoud
ABSTRACT
We investigated whether neonatal exposure to low doses of endosulfan affects fertility and uterine functional differentiation at pre-implantation in rats. Newborn female rats received the vehicle, 0.2 µg/kg/d of diethylstilbestrol (DES), 6 µg/kg/d of endosulfan (Endo6) or 600 µg/kg/d of endosulfan (Endo600) on postnatal days (PND) 1, 3, 5, and 7. On PND90, the rats were mated to evaluate their reproductive performance on gestational day (GD) 19 and their ovarian steroid serum levels, endometrial proliferation and implantation-associated proteins on GD5. DES and endosulfan decreased the pregnancy rate and the number of implantation sites. On GD5, DES and endosulfan did not change the serum levels of 17β-estradiol (E2) and progesterone (P); the endometrial proliferation decreased, which was associated with silencing of Hoxa10 in the Endo600-treated rats. Both doses of endosulfan increased the progesterone receptor (PR) expression, whereas the higher dose led additionally to an increase in estrogen receptor alpha (ERα). In the Endo600-treated rats, the down-regulation of Hoxa10 was associated with a deregulation of the steroid receptor coregulators. Alterations in endometrial proliferation and the endocrine pathway of Hoxa10/steroid receptors/coregulators might be the mechanism of endosulfan-induced implantation failure.
Product Description
Supelco
Endosulfan, PESTANAL®, analytical standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free