Skip to Content
Merck
All Photos(1)

Documents

ERMBF445B

DP23211 Maize

nominal 100% GMO, ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

product name

DP23211 Maize (nominal 100% GMO),

manufacturer/tradename

JRC

application(s)

food and beverages

storage temp.

2-8°C

Analysis Note

For more information please click on the below product number:
ERMBF445B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service