Skip to Content
Merck
All Photos(1)

Key Documents

821971

TSKgel® NH2-100 (3 µm) HPLC Columns

L x I.D. 1 cm × 2 mm, amino phase,amine phase

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
4 160,00 kr

4 160,00 kr


Please contact Customer Service for Availability


Select a Size

Change View
3 EA
4 160,00 kr

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

4 160,00 kr


Please contact Customer Service for Availability

Product Name

TSKgel® NH2-100 Guard Cartridge, phase amine, L × I.D. 1 cm × 2 mm, 3 μm particle size, pkg of 3 ea

material

stainless steel

Agency

suitable for USP L8

product line

TSKgel®

packaging

pkg of 3 ea

manufacturer/tradename

Tosoh 21971

parameter

0-100% organic solvent
10-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1 cm × 2 mm

surface area

450 m2/g

matrix

silica particle platform

matrix active group

amine phase
amino phase

particle size

3 μm

pore size

100 Å

operating pH

2.0-7.5

separation technique

anion exchange
hydrophilic interaction (HILIC)
normal phase

Looking for similar products? Visit Product Comparison Guide

General description

Guard Cartridge for 2mmID TSKgel NH2-100 columns, 3μm, 3pk

Physical form

Shipped in 85% acetonitrile - 15% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

required but not provided

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service