Skip to Content
Merck
All Photos(1)

Documents

Z691992

Nunc® Lab-Tek® Chamber Slide system, 4 wells on glass

working volume 0.5-0.9 μL

Synonym(s):

cell culture chamber

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

material

colorless
glass

sterility

sterile

feature

CE compliant
wells 4

packaging

pkg of 96 ea

manufacturer/tradename

Nunc 177399

W × L

25 mm × 75 mm

well area

1.8 cm2

wells

4

working volume

0.5-0.9 μL

General description

  • Cells grow on a standard microscope slide
  • No cell transfer needed prior to visualisation/staining
  • Upper structure can be removed when culturing is complete
  • Useful for viral and mycoplasma testing, chromosome studies, toxicity tests and immunocytology
  • Broad range of formats and well numbers
  • Fits standard equipment
  • Saves time and reagents
  • Suitable for use with fluorescent labels

Legal Information

Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service