Key Documents
Z367885
Scienceware® colony counter system
Magnifier for mini light box, free-standing
About This Item
UNSPSC Code:
41100000
manufacturer/tradename
Bel-Art
General description
This free standing magnifier allows enhanced viewing of slides, petri dishes, multi-well plates, gels or text. The precision molded lens offers 1.75 magnification. The unique stand allows the magnifier to be placed over a variety of objects, including the Mini Light Box. Dimensions: lens 104.8 x 142.9 mm (41/8x 55/8 in.), stand 171.8mm (6 3/4 in.) high.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)