Skip to Content
Merck
All Photos(1)

Documents

V5390

Sigma-Aldrich

VU0029251

≥98% (HPLC), powder

Synonym(s):

6,7-Dihydro-2-(methylthio)-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N3S2
Molecular Weight:
237.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥11 mg/mL

storage temp.

2-8°C

SMILES string

CSc1nc(N)c2c3CCCc3sc2n1

InChI

1S/C10H11N3S2/c1-14-10-12-8(11)7-5-3-2-4-6(5)15-9(7)13-10/h2-4H2,1H3,(H2,11,12,13)

InChI key

WDDBZCKBRCTSOZ-UHFFFAOYSA-N

Biochem/physiol Actions

VU0029251 is the first example of a partial mGluR5 partial antagonist that is not based on the MPEP scaffold. The compound displays a maximum inhibition of 50% activity, and cannot completely displace [3H] MPEP, suggesting that competition for MPEP is also allosteric, involving a unique site.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service