IST121080TS
QuickSeal qPCR Crystal™ self adhesive sealing film
LabSheet™, W × L 80 mm × 140 mm, pkg of pack of 5 sheets (trial pack)
Synonym(s):
Self adhesive seal, plate sealing film, pressure activated seal
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
sterility
non-sterile
manufacturer/tradename
IST Scientific IST-121-080TS
packaging
pkg of pack of 5 sheets (trial pack)
technique(s)
qPCR: suitable
W × L
80 mm × 140 mm
suitability
suitable for fluorescence
Looking for similar products? Visit Product Comparison Guide
General description
Optically clear self adhesive film with pressure activated adhesive. The adhesive is contained within small capsules, allowing light to pass through to ensure the optical clarity of the seal. When the seal is in position, pressure can be applied to burst the capsules, releasing a strong adhesive only where the seal touches the raised well rims of the plate – the rest of the seal area above the wells remains optically clear.
Application
qPCR and other fluorescent applications, plate readers, microscopy and protein crystallization (96 or 384 well)
Features and Benefits
- Non-Pierceable
- Peelable
- Seal integrity range: -80 °C to 110 °C
- Will not leave a sticky residue on the plate surface following removal
Legal Information
ClearASeal Peel is a trademark of IST Scientific
LabSheet is a trademark of IST Scientific
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service