EP5810711002
Eppendorf® Swing-bucket rotor A-4-62-MTP
A-4-62-MTP with 4 MTP buckets
All Photos(1)
About This Item
Recommended Products
feature
to hold 4 x MTP buckets
A-4-62-MTP with 4 MTP buckets
packaging
pack of 1 ea
manufacturer/tradename
5810711002
Eppendorf® 5810711002
parameter
4,000 max. RCF (3,220 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
General description
- Eppendorf® Swing-bucket rotor accommodates tubes and bottles from 0.2 mL to 250 mL as well as plates.
- Aerosol-tight caps for safe centrifugation of hazardous samples optionally available.
- Rotor life time: max 40,000 cycles.
- Rotor, buckets, caps and adapters are autoclavable.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG