Skip to Content
Merck
All Photos(1)

Key Documents

B1D6

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 98042014-DNA-5UG
pkg of vial of cells 98042014-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

P3X63Ag8

Reactivity

Anti-annexin II, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Not specified

Cell Line Description

The hybridoma B1D6 was obtained by immunising BALB/c mice with human placental extract followed by intravenous injections with purified Fc gamma-receptor (receptor for the Fc portion of immunoglobulins, FcR). Spleen cells were then fused with the myeloma P3X63Ag8 to establish B1D6 secreting IgG1, kappa, monoclonal antibodies. This antibody has been shown to stain FcR-positive areas in sections of placental tissues, inhibited the haemadsorption of placental tissue of erythrocytes sensitised with IgG antibodies and the agglutination of these erythrocytes by FcR.

Immunogen

Human placenta FcR

Application

FcR studies

Culture Medium

RPMI 1640 + 2 mM glutamine + 10% FBS

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37 °C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service