Skip to Content
Merck
All Photos(1)

Key Documents

79279

Sigma-Aldrich

3-Phenylumbelliferone

suitable for fluorescence, ≥98.0% (TLC)

Synonym(s):

7-Hydroxy-3-phenylcoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O3
CAS Number:
Molecular Weight:
238.24
Beilstein:
384125
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98.0% (TLC)

form

crystals

pKa 

7.8

mp

206-210 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
alcohols: soluble

fluorescence

λex 338 nm; λem 463 nm (pH 3.0)
λex 383 nm; λem 472 nm in 0.1 M Tris pH 9.0

suitability

suitable for fluorescence

SMILES string

Oc1ccc2C=C(C(=O)Oc2c1)c3ccccc3

InChI

1S/C15H10O3/c16-12-7-6-11-8-13(10-4-2-1-3-5-10)15(17)18-14(11)9-12/h1-9,16H

InChI key

RIPZCQZTVDNJHQ-UHFFFAOYSA-N

Gene Information

human ... MIF(4282)

Application

suitable as pH-indicator

Other Notes

Fluorescent pH probe for the physiological pH range[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O.S. Wolfbeis
Zeitschrift fur Naturforschung, A, 32, 1065-1065 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service