Skip to Content
Merck
All Photos(1)

Key Documents

31755

Sigma-Aldrich

7-(Diethylamino)coumarin-3-carbonyl azide

BioReagent, suitable for fluorescence, ≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4O3
CAS Number:
Molecular Weight:
286.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

product line

BioReagent

Assay

≥95% (HPLC)

form

solid

fluorescence

λex 390 nm; λem 478 nm in methanol

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CCN(CC)c1ccc2C=C(C(=O)Oc2c1)C(=O)N=[N+]=[N-]

InChI

1S/C14H14N4O3/c1-3-18(4-2)10-6-5-9-7-11(13(19)16-17-15)14(20)21-12(9)8-10/h5-8H,3-4H2,1-2H3

InChI key

FJIQMYGHCNXCHE-UHFFFAOYSA-N

Other Notes

Fluorescent labeling of 2-dodecylcyclobutanone[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.-T. Morsel et al.
Fresenius Journal of Analytical Chemistry, 349, 538-538 (1994)
Tomáš Urbánek et al.
Macromolecular bioscience, 20(5), e1900408-e1900408 (2020-03-17)
Polymeric drug carriers exhibit excellent properties that advance drug delivery systems. In particular, carriers based on poly(ethylene oxide)-block-poly(ε-caprolactone) are very useful in pharmacokinetics. In addition to their proven biocompatibility, there are several requirements for the efficacy of the polymeric drug
Christopher J Keturakis et al.
ChemSusChem, 11(23), 4044-4059 (2018-10-20)
Biomass pyrolysis by solid acid catalysts is one of many promising technologies for sustainable production of hydrocarbon liquid fuels and value-added chemicals, but these complex chemical transformations are still poorly understood. A series of well-defined model SiO2 -supported alumina catalysts
Markus Blaess et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 986-987, 123-128 (2015-03-03)
Ceramides are derivatised using 7-(diethylamino)coumarin-3-carbonyl azide; subsequent gradient HPLC separation allows sensitive optical quantification of individual cellular ceramides. Compared to 9-anthracenecarbonyl cyanide (9-anthroyl nitrile) as derivatisation agent, the limit of detection could be improved 415-fold, respectively 10,000-fold (detection limit 0.6

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service