Skip to Content
Merck
All Photos(1)

Documents

Y0001017

Methylphenidate hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

threo-Methyl α-phenyl-α-(2-piperidyl)acetate hydrochloride, Ritalin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO2 · HCl
CAS Number:
Molecular Weight:
269.77
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methylphenidate

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].COC(=O)C(C1CCCCN1)c2ccccc2

InChI

1S/C14H19NO2.ClH/c1-17-14(16)13(11-7-3-2-4-8-11)12-9-5-6-10-15-12;/h2-4,7-8,12-13,15H,5-6,9-10H2,1H3;1H

InChI key

JUMYIBMBTDDLNG-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylphenidate hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Britahny M Baskin et al.
Pharmacology, biochemistry, and behavior, 131, 51-56 (2015-02-04)
Past research with the spontaneously hypertensive rat (SHR) model of attention deficit/hyperactivity disorder showed that adolescent methylphenidate treatment enhanced cocaine abuse risk in SHR during adulthood. The acquisition of cocaine self-administration was faster, and cocaine dose-response functions were shifted upward
Panayotis K Thanos et al.
Pharmacology, biochemistry, and behavior, 131, 143-153 (2015-02-03)
Most animal studies using methylphenidate (MP) do not administer it the same way it is administered clinically (orally), but rather by injection, resulting in an altered pharmacokinetic profile (quicker and higher peak concentrations). We evaluated several oral-dosing regimens in rats
Chloe J Jordan et al.
Drug and alcohol dependence, 140, 25-32 (2014-05-09)
Attention-deficit/hyperactivity disorder (ADHD) is often comorbid with cocaine abuse. Controversy exists regarding long-term consequences of ADHD medications on cocaine abuse liability. Whereas childhood methylphenidate treatment may be preventative, methylphenidate in teens appears to further increase later cocaine abuse risk. In
Craige C Wrenn et al.
Neuroscience letters, 585, 109-113 (2014-11-30)
Fragile X syndrome (FXS), a disorder caused by a mutation in the FMR1 gene, is often associated with Attention Deficit Hyperactivity Disorder (ADHD). Common treatments for the hyperactivity often seen in ADHD involve the use of stimulants and α2-adrenergic agonists.
Alexander Frolov et al.
Journal of neurophysiology, 113(1), 369-379 (2014-10-17)
The nucleus accumbens (NAc) has been shown to play a key role in the brain's response to methylphenidate (MPD). The present study focuses on neuronal recording from this structure. The study postulates that repetitive exposure to the same dose of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service