Recommended Products
grade
certified reference material
CofA
current certificate can be downloaded
packaging
pkg of 50 g
manufacturer/tradename
RTC CRM141-050
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
matrix
soil
application(s)
environmental
format
matrix material
storage temp.
2-8°C
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Analyte
Description
Acenaphthene
Acenaphthylene
Aldrin
Anthracene
Benzo[b]fluoranthene
Benzo[k]fluoranthene
Benzo[ghi]perylene
Benzo[a]pyrene
Chrysene
1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane
2,4′-DDE
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene
2,4′-DDT
4,4′-DDT
Dibenz[a,h]anthracene
Dieldrin
α-Endosulfan
Endrin
Fluoranthene
Fluorene
α-HCH
β-HCH
Heptachlor
2,2′,3,4,4′,5,5′-Heptachlorobiphenyl
2,2′,3,4,4′,5′-Hexachlorobiphenyl
2,2′,4,4′,5,5′-Hexachlorobiphenyl
Naphthalene
2,2′,4,5,5′-Pentachlorobiphenyl
2,3′,4,4′,5-Pentachlorobiphenyl
Phenanthrene
2,2′,5,5′-Tetrachlorobiphenyl
2,4,4′-Trichlorobiphenyl
See All (33)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service