Skip to Content
Merck
All Photos(1)

Documents

92659

Supelco

4-[2-[Benzyl(tert-butyl)amino]-1-hydroxyethyl]-2-(hydroxymethyl)phenol

pharmaceutical impurity standard

Synonym(s):

Salbutamol impurity E (PhEur), α1-[[(1,1-Dimethylethyl)(phenylmethyl)amino]methyl]-4-hydroxy-1,3-benzenedimethanol, (1RS)-2-[Benzyl(1,1-dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27NO3
CAS Number:
Molecular Weight:
329.43
EC Number:
MDL number:
UNSPSC Code:
41116107

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

InChI

1S/C20H27NO3/c1-20(2,3)21(12-15-7-5-4-6-8-15)13-19(24)16-9-10-18(23)17(11-16)14-22/h4-11,19,22-24H,12-14H2,1-3H3

InChI key

NZBXAYHLQSMNQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service