Skip to Content
Merck
All Photos(1)

Documents

84700

Sigma-Aldrich

Sulfur hexafluoride

purum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SF6
CAS Number:
Molecular Weight:
146.06
EC Number:
MDL number:
UNSPSC Code:
12352302

vapor density

5.11 (vs air)

vapor pressure

22 mmHg ( 21.1 °C)

grade

purum

impurities

0.001% S2F10
0.002% SiF4
1-2% Air

bp

−64 °C/1 mmHg (lit.)

mp

−50 °C (lit.)

SMILES string

FS(F)(F)(F)(F)F

InChI

1S/F6S/c1-7(2,3,4,5)6

InChI key

SFZCNBIFKDRMGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Cylinder with net ~4.75 kg

Other Notes

Sales restrictions may apply

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service