Skip to Content
Merck
All Photos(1)

Key Documents

46917

Supelco

9-Fluorenylmethyl carbazate

for HPLC derivatization, LiChropur, ≥99.0%

Synonym(s):

Fmoc-hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2O2
CAS Number:
Molecular Weight:
254.28
Beilstein:
2470262
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

grade

for HPLC derivatization

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

quality

LiChropur

technique(s)

HPLC: suitable

mp

~170 °C (dec.)

fluorescence

λex 264 nm; λem 309 nm in methanol
λex 270 nm; λem 320 nm (Saccharide adducts)

storage temp.

2-8°C

SMILES string

NNC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C15H14N2O2/c16-17-15(18)19-9-14-12-7-3-1-5-10(12)11-6-2-4-8-13(11)14/h1-8,14H,9,16H2,(H,17,18)

Application

9-Fluorenylmethyl carbazate may be used as a derivatization reagent to derivatize progesterone for high-performance liquid chromatography, in the analysis of progesterone, 17-hydroxyprogesterone and other 3-keto steroids.[1]It may also be used as one of the fluorescence labeling agent for high-performance liquid chromatography determination of neutral and amino monosaccharides in glycoproteins by ultraviolet and fluorescence detection.[2]

Other Notes

Reagent for the sensitive fluorogenic derivatization of carbohydrates[3][4]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

High-performance liquid chromatographic determination of neutral and amino monosaccharides by ultraviolet and fluorescence detection of sugar 9-fluoroenylmethoxycarbonyl hydrazones and 9-fluoroenylmethoxycarbonyl amino sugars at picomole and sub-picomole levels.
Zhang R, et al.
Journal of Chromatography A, 646(1), 45-52 (1993)
Determination of progesterone and 17-hydroxyprogesterone by high performance liquid chromatography after pre-column derivatization with 4, 4-difluoro-5, 7-dimethyl-4-bora-3a, 4a-diaza-s-indacene-3-propionohydrazide.
Katayama M, et al.
Analyst, 123(11), 2339-2342 (1998)
Jinmao You et al.
Analytica chimica acta, 636(1), 95-104 (2009-02-24)
A pre-column derivatization method for the sensitive determination of aldehydes using the tagging reagent 2-[2-(7H-dibenzo[a,g] carbazol-7-yl)-ethoxy] ethyl carbonylhydrazine (DBCEEC) followed by high-performance liquid chromatography with fluorescence detection and APCI-MS identification has been developed. The chromophore of fluoren-9-methoxy-carbonylhydrazine (Fmoc-hydrazine) reagent was
R E Zhang et al.
Analytical biochemistry, 195(1), 160-167 (1991-05-15)
A new derivatization reagent, Fmoc-hydrazine, has been synthesized from the reaction of Fmoc-chloroformate with hydrazine as a precolumn fluorometric labeling reagent for reducing sugars such as glucose, galactose, mannose, fructose, fucose, ribose, xylose, arabinose, lactose, and maltose. The optimization of
Journal of Chromatography A, 646, 45-45 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service