39833
3,3-Dimethyl-1-butene
analytical standard
Synonym(s):
Neohexene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
analytical standard
vapor density
>1 (vs air)
vapor pressure
6.96 psi ( 20 °C)
Assay
≥99.5% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.376 (lit.)
n20/D 1.377
bp
41 °C (lit.)
mp
−115 °C (lit.)
density
0.653 g/mL at 25 °C (lit.)
application(s)
environmental
petroleum
SMILES string
CC(C)(C)C=C
InChI
1S/C6H12/c1-5-6(2,3)4/h5H,1H2,2-4H3
InChI key
PKXHXOTZMFCXSH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service